Would you like to log in again?
Choice® NW 600 Non-Woven Cleanroom Wipes, Berkshire
Supplier: Berkshire
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CHOICE®
CHNW60440 CHNW60920 CHNW61220
76077-274CS 418.59 USD InStock
76077-274 76077-514 76077-516
Choice® NW 600 Non-Woven Cleanroom Wipes, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Choice® NW 600 wipes are composed of a hydroentangled nonwoven blend of 55% cellulose and 45% polyester.
  • Excellent combination of synthetic polyester strength and cleanliness
  • Chemically compatible with common cleaning and disinfecting solutions
  • Autoclavable
  • Economical
They offer all the performance consistency of higher priced wipes at an economical price. These wipes are a level of absorbency, abrasion-resistance, and chemical compatibility required for controlled environments.
They are designed for use in ISO Class 5 and higher cleanroom environments.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty