Would you like to log in again?
Biox Tissue Culture Flasks
Supplier: TradeWinds (TWD)
Biox
07-8025 BX07-9025 BX07-9075 07-8075
76081-542CS 300.63 USD InStock
76081-542 76085-626 76081-544 76081-540
Biox Tissue Culture Flasks
Flasks Cell Culture Flasks
High quality polystyrene cell culture flasks are sterile, stackable, and a great value.
  • High-quality medical grade polystyrene
  • Plug caps completely isolate the culture from the outside environment
  • Filter Caps prevent cross-contamination while meeting the requirements for gas exchange
  • Electron beam sterilized
  • Short, wide, angled neck design for easy access
  • Non-pyrogenic and non-cytotoxic
  • DNase- & RNase-free, Human DNA-free
TC treated cell culture flasks with plug or filter caps allow for complete isolation from the outside environment and prevent cross contamination while allowing for gas exchange.
Certifications: ISO Certified: GB/T 19001-2008/ISO 9001:2008.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty