Would you like to log in again?
Biox Culture Dishes
Supplier: TradeWinds (TWD)
Biox
BX07-3035 BX07-3060 BX07-3150 BX07-3100
76081-548CS 278.36 USD InStock
76081-548 76081-554 76081-550 76081-552
Biox Culture Dishes
Culture Dishes Cell Culture Dishes
Sterile and constructed of optically clear material, these cell culture dishes offer high quality at a great price.
  • High-quality medical grade polystyrene
  • Vacuum plasma surface treatment ensures excellent cell adhesion
  • Lid design promotes effective gas exchange
  • External grip for easier handling (available on 2 models)
  • Electron beam sterilized
  • Non-pyrogenic and non-cytotoxic
  • Dnase- & Rnase-free, human DNA-free
These TC treated culture dishes have vented lids and raised edges for convenient stacking.
Certifications: ISO Certified: GB/T 19001-2008/ISO 9001:2008.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty