Would you like to log in again?
MasterPure™ Bulk Reagents, Reagents for Nucleic Acid Purification, Biosearch Technologies
Supplier: Biosearch Technologies
MasterPure™
MTE0970 MMP095H MRC0912H MTC096H MPRK092
76081-700EA 632.82 USD InStock
76081-700 76081-702 76081-696 76081-644 76081-660
MasterPure™ Bulk Reagents, Reagents for Nucleic Acid Purification, Biosearch Technologies
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Save by purchasing bulk quantities of the MasterPure™ buffers and reagents
  • Save: Reduce costs by purchasing these important reagents in bulk quantities
  • Scale-up: Use these large volumes of reagents to perform large-scale MasterPure™ purifications
  • Adapt: Use these reagents whenever your experiments require these high quality reagents
Proteinase K: Useful for multiple applications outside of the MasterPure™ Kits including (i) isolation of plasmids and high molecular weight gDNA, purification of intact RNA, and inactivation of RNases, DNases and other enzymes in molecular biology reactions
Red Cell Lysis Solution: Selectively lyse red blood cells while leaving white blood cells intact
Tissue and Cell Lysis Solution: Use in combination with Proteinase K to lyse cells and tissues in various samples such as saliva, semen, mammalian cells, and gram negative bacteria
Certifications: For research use only. Not for human or diagnostic use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty