Would you like to log in again?
Orion™ Navigator Pro Software, Thermo Scientific
Supplier: Thermo Scientific
VSTAR-NPCD VSTAR-NPUSB
76085-810EA 862.88 USD InStock
76085-810 76085-812
Orion™ Navigator Pro Software, Thermo Scientific
Software Laboratory Software
Ensure digital data integrity with password-protection, electronic records, and audit trails.
  • Generate electronic records and signatures
  • Create audit trail report for electronic records
  • Limits access to authorized users
  • Password protection with system administrator and up to 10 users
  • Full computer control of meter functions including setup, calibration and measurement
  • Paperless record keeping system
  • Memory: 2 GB
  • Display resolution: 1366×768 true color
  • Compatible with Microsoft Windows 7, 8.1, 10 operating systems
  • CPU Type: 1 GHz or faster 32 bit (x86) or 64 bit (x64) processor
The Orion™ Navigator Pro computer software paired with the Orion™ Versa Star Pro™ meters are designed to offer a computer-driven measurement system for generating electronic measurements, calibration, and audit trails.
Orion™ Navigator Pro software offers multi-level system access for an administrator and up to 10 users. The system administrator can customize each user's access level from full system access to basic data collection only. The Navigator Pro software controls all meter functions inlcuding meter setting, calibration, measurement, and reporting to help ensure data integrity and provide a complete audit trail.
Orion™ Navigator Pro software is designed to create electronic data records and audit trails with password protection, which can assist with 21 CFR Part 11 requirements when paired with each laboratory's proper IT protocols, SOP's, and training records.
Caution: Note: It is the responsibility of each laboratory to validate the Navigator Pro computer software for its intended use, including all regulatory requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty