Would you like to log in again?
Gamma Wipe® 120 Cleanroom Laundered Wipes, Berkshire
Supplier: Berkshire
Gamma Wipe®
GW120ST15 GW120ST0940
21913-811CS 832.15 USD InStock
21913-811 76097-928
Gamma Wipe® 120 Cleanroom Laundered Wipes, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Each package of Gamma Wipe® 120 wipes is gamma irradiated sterile to a 10⁻⁶ sterility assurance level. This combination of properties provides them with the high level of cleanliness, abrasion resistance, and chemical compatibility required for cleaning critical sterile processing environments.
  • Low particles, fibers, ions, and extractables
  • High absorbency
  • High abrasion resistance
  • Soft hand and edge for sensitive surfaces
  • Chemically compatible with common cleaning and disinfecting solutions, including isopropyl alcohol, phenols, bleaches, and quarternary ammonium compounds
Wipes are a knife-cut, high absorbency cleanroom laundered wiper recommended for ISO Class 4 and above environments. The wipes are composed of 100% continuous filament polyester knit fabric.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty