Would you like to log in again?
Excellence XPR Legal for Trade (NTEP) Precision Balances, METTTLER TOLEDO®
Supplier: Mettler-Toledo
30317452 30317126 30317436 30317486 30315651 30317404 30317494 30317500 30317442 30316480 30316528 30315627 30316566 30316543 30316508 30317461
76098-090EA 21480 USD InStock
76098-090 76098-126 76098-028 76098-128 76098-122 76098-100 76098-134 76098-102 76098-124 76098-136 76098-114 76098-140 76098-030 76098-120 76098-032 76098-132
Excellence XPR Legal for Trade (NTEP) Precision Balances, METTTLER TOLEDO®
Balances and Scales
XPR balances deliver accurate results, outstanding weighing performance and support the highest requirements for data integrity and compliance. Quality assurance features ensure your weighing results are always valid, helping you to comply with even the strictest regulations.
  • Made of chemically resistant materials
  • 7" color capacitive touchscreen terminal, glove compatible
  • Storage of weighing methods (including sample series and tolerances)
  • Levelcontrol: level warning with graphical leveling guide
  • Easy connectivity and data export: 4×USB, 1 LAN
  • Integrated label editor, alphanumeric and barcode
  • Export of process and results data to XML or Excel
  • Integrated overload protection
  • Designed for easy cleaning
LabX automatically saves all results and process information securely in a central database. This ensures traceability and assists with FDA 21 CFR part 11 compliance.
The extraordinary stability provided by the new Monobloc™ weighing cell as well as the rugged balance design means you get your results fast even under challenging production environments.
Offer easy connection of peripheral devices like printers, PC, barcode readers, footswitches and sensors. Designed with temperature and time programmable automatic adjustment (proFACT). With customizable tolerance profiles and minimum weigh warning function.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty