Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magne-Box™ Magnetic Polycarbonate Cryoboxes, Argos Technologies

Supplier: Argos Technologies

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Magne-Box™
04200-06 04200-04 04200-05 04200-03
76102-172PK 219.04 USD InStock
76102-172 76102-170 76102-174 76102-168
Magne-Box™ Magnetic Polycarbonate Cryoboxes, Argos Technologies
Boxes Cryoboxes
Magne-Box™ freezer boxes are the first of its kind to offer a magnetic lid closure.

  • Boxes fit in standard size freezer racks
  • Polycarbonate design withstands temperatures from –196 to 121 °C
  • Slot openings on the bottom of boxes allow for drainage
  • 25-, 81-, and 100-Place boxes accommodate 2 mL cryovials
  • 81-Place Tall boxes accommodate 4 to 5 mL cryovials

Each box features magnets that securely connect the base to the hinged lid. This allows one-handed retrieval from freezer racks and ensures that samples stay safe. Polycarbonate boxes have clear lids with colored inserts and numbered grids to facilitate easy sample identification. Available with 25-, 81-, and 100-place divisions.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty