Would you like to log in again?
Polka Dot Grip Gloves, Portwest
Supplier: Portwest
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
A110WBRXL A110WBRL A110WBRM
76103-330EA 1.89 USD InStock
76103-330 76103-332 76103-334
Polka Dot Grip Gloves, Portwest
Gloves
By combining enhanced grip with excellent dexterity, the Polka Dot Glove performs well in dry conditions.
  • Seamless 13 gauge knitted liner with PVC dotted palm
  • Extra coating on the fingertips to improve small item pick up
  • Performs well in dry conditions
Excellent for jobs requiring high dexterity
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. ANSI/ISEA 105 - 2016 ABRASION Level 1. EN 388:2016 - 2.1.4.X.X. EN 420.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty