Would you like to log in again?
PU Palm Gloves, Portwest
Supplier: Portwest
A120GRRXXL A120BKRXS A120WHRXL A120BKRXL A120WHRS A120BKRXXL A120BKRS A120GRRL A120BKRM A120Y2RL A120GRRS A120O1RXL A120WHRXXL A120O1RL A120WHRM A120B4RM A120O1RS A120WHRXS A120P9RXS A120GRRXS A120WHRL A120B4RL A120GRRM A120BKRL A120O1RM A120P9RS A120GRRXL A120P9RM A120Y2RXL A120Y2RM A120B4RXL
76103-380EA 1.56 USD InStock
76103-380 76103-382 76103-360 76103-384 76103-362 76103-340 76103-364 76103-386 76103-342 76103-344 76103-366 76103-388 76103-346 76103-368 76103-348 76103-338 76103-390 76103-392 76103-370 76103-350 76103-372 76103-394 76103-352 76103-374 76103-396 76103-354 76103-376 76103-378 76103-356 76103-358 76103-336
PU Palm Gloves, Portwest
Gloves
Offering maximum dexterity, this glove is perfect for intricate tasks.
  • Smooth polyurethane coating for increased abrasion resistance
  • Palm dipped to increase dexterity and ventilation
  • Superb tear resistance
  • 100% breathable seamless pylon liner
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. ANSI/ISEA 105 - 2016 CUT Level A1. EN 388:2016 - 2.0.3.1.X. EN 388:2003 - 4.1.3.1.EN 420.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty