Would you like to log in again?
Antistatic PU Fingertip Gloves, Portwest
Supplier: Portwest
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
A198GRRXS A198GRRXXL A198GRRS A198GRRM A198GRRL A198GRRXXS
76103-504EA 1.99 USD InStock
76103-504 76103-508 76103-500 76103-510 76103-502 76103-512
Antistatic PU Fingertip Gloves, Portwest
Gloves
Antistatic PU fingertip Gloves are made for use in electronic assembly, testing and precision work. Special construction diverts static electricity.
  • Pylon and carbon fiber liner material
  • 13 gauge pylon and carbon fiber shell
  • Smooth PU coating for increased abrasion resistance
  • Fingertip dipped for precision handling and maximum ventilation
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. ANSI/ISEA 105 - 2016 CUT Level (A1). EN 420. EN1149. EN 388:2003 - 2.1.3.2. EN 388:2016 - 1.1.4.X.X.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty