Would you like to log in again?
Dermiflex Ultra Gloves, Portwest
Supplier: Portwest
A352G8RXL A352G8RM A352G8RL
76103-614EA 4.75 USD InStock
76103-614 76103-610 76103-612
Dermiflex Ultra Gloves, Portwest
Gloves
Offering superb abrasion and tear resistance, plus an excellent grip in both wet and dry conditions.
  • Maximum dexterity
  • PU/nitrile blended foam coating
  • ³/₄ dipped for increased protection
  • Open back for breathability
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. ANSI/ISEA 105 - 2016 CUT Level (A1). EN3 88:2003 - 3.1.3.1. EN 388:2016 - 4.1.3.1.X. EN 420.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty