Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
MR Cut PU Palm Gloves, Portwest
Supplier: Portwest
A622G7RXXL A622G7RM A622G7RXL A622G7RL
76103-650EA 7.67 USD InStock
76103-650 76103-652 76103-654 76103-656
MR Cut PU Palm Gloves, Portwest
Gloves
Level C cut resistant glove for protection against cuts and sharp objects. Seamless liner for added comfort and dexterity.
  • Level C cut resistance
  • 13 gauge liner for a perfect fit
  • Smooth PU coating for increased abrasion resistance
  • Palm dipped to increase dexterity and ventilation
  • Breathable seamless liner
  • Medium risk cut protection
  • Superb abrasion and tear resistance
EN 407 contact heat level 1 provides short term protection against temperatures up to 100 °C.
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. AS/NZS 2161 .4. EN 388:2016 - 4.X.4.3.C. EN 407 X.1.X.X.X.X. ANSI/ISEA 105 - 2011 Cut Level (A2). EN 420. EN 388:2003 - 4.5.4.3. ANSI cut level A3.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty