Would you like to log in again?
Anti Vibration Gloves, Portwest
Supplier: Portwest
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
A790BKRXL A790BKRM A790BKRL
76103-704EA 32.57 USD InStock
76103-704 76103-706 76103-708
Anti Vibration Gloves, Portwest
Gloves
Specially designed to reduce the effects of impact and vibration when using jack hammers, concrete breakers etc.
  • 100% breathable seamless cotton/nylon liner
Anti-vibration pods covering palms are made of specially formulated rubber chloroprene.
Certifications: AS/NZS 2161 .2. AS/NZS 2161 .3. EN388:2016 - 3.1.4.2.X. EN420. EN10819 (TRm=0.62 TRh=0.55) vibration and shock protection. Meets anti-vibration glove standards EN 10819.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty