Would you like to log in again?
Compositelite™ Operis FC60, Safety Boots, Lace-Up, Portwest
Supplier: Portwest
Compositelite™ Operis
FC60BKR44 FC60BKR37 FC60BKR41 FC60BKR42 FC60BKR45 FC60BKR40 FC60BKR38 FC60BKR46 FC60BKR47 FC60BKR48 FC60BKR39
76103-944EA 134.96 USD InStock
76103-944 76103-946 76104-058 76104-046 76104-056 76104-044 76104-054 76104-052 76104-050 76103-940 76103-942
Compositelite™ Operis FC60, Safety Boots, Lace-Up, Portwest
Shoes and Boots
Stylish design combined with total S3 safety in a 100% non metallic boot made fom nubuck leather.
  • Protective composite toecap
  • Puncture-resistant composite midsole
  • Lightweight PU cushioned midsole
  • Slip-, fuel-, and oil-resistant PU/rubber outsole
  • Direct injected dual density construction
  • Heat-resistant outsole
  • 100% metal-free
  • Anti-static
Lightweight construction with a durable PU/Rubber outsole for slip resistance and shock absorption.
Color: Black
Certifications: ASTM F2413-11
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty