Would you like to log in again?
Steelite™ Protector FW14, Safety Shoes, Lace-Up, Portwest
Supplier: Portwest
Steelite™
FW14BKR36 FW14BKR46 FW14BKR40 FW14BKR39 FW14BKR51 FW14BKR47 FW14BKR42 FW14BKR49 FW14BKR41 FW14BKR44 FW14BKR37 FW14BKR38 FW14BKR48 FW14BKR50 FW14BKR43 FW14BKR35 FW14BKR52 FW14BKR45
76104-402EA 50.33 USD InStock
76104-402 76104-412 76104-148 76104-158 76104-400 76104-410 76104-156 76104-398 76104-154 76104-396 76104-152 76104-162 76104-150 76104-160 76104-408 76104-406 76104-404 76104-414
Steelite™ Protector FW14, Safety Shoes, Lace-Up, Portwest
Shoes and Boots
This popular safety shoe is lightweight and durable, allowing wearers to work confidently in any environment. It is made from split buffalo leather and oil resistant PU/PU outsole.
  • Protective steel toecap
  • Puncture-resistant steel midsole
  • Direct injected dual density construction
  • Lightweight PU cushioned midsole
  • Slip-, fuel-, and oil-resistant outsole
  • Removable EVA cushion footbed
  • Anti-static
Superb slip-resistant, oil-resistant, and energy absorbing outsole with protective steel toecap and midsole.
Color: Black
Certifications: ASTM F2413-11
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty