Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Steelite™ Total FW95, Safety Wellington Boots, Portwest

Supplier: Portwest

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Steelite™ Total
FW95BKR37 FW95BKR46 FW95BKR38 FW95BKR41 FW95BKR48 FW95BKR39 FW95BKR40 FW95BKR45 FW95BKR43 FW95BKR47 FW95BKR49 FW95BKR42 FW95BKR36 FW95BKR44
76104-864EA 52.3 USD InStock
76104-864 76104-468 76104-478 76104-488 76104-862 76104-476 76104-486 76104-474 76104-484 76104-472 76104-482 76104-470 76104-480 76104-866
Steelite™ Total FW95, Safety Wellington Boots, Portwest
Shoes and Boots
The Total Safety Wellington offers ultimate S5 protection to the wearer

  • Protective steel toecap
  • Puncture-resistant steel midsole
  • Dual density PVC/Nitrile construction
  • Slip-, fuel-, and oil-resistant PVC/Nitrile outsole
  • 100% waterproof
  • Washable nylon inner lining
  • 15" height
  • Anti-static

Steel toecap and midsole. PVC/Nitrile construction that is waterproof and resistant to oil, fuels, and acids. Suitable for a variety of environments.

Color: Black

Certifications: ASTM F2413-11

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty