Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

High Visibility 48" Classic Rain Coats, Portwest

Supplier: Portwest

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

UH445YERM UH445YERXL UH445YERL UH445YERXXL UH445YER5XL UH445YER4XL UH445YERXXXL
76105-950EA 61.91 USD InStock
76105-950 76105-946 76105-956 76105-948 76105-952 76105-944 76105-954
High Visibility 48" Classic Rain Coats, Portwest
Workwear
Waterproof with sealed seams, mic tab, and badge holder.

  • Pack-away adjustable hood
  • Storm flap and zipper closure
  • 2" silver reflective tape

These are designed with two deep front pockets and elasticated storm cuffs.

Certifications: ANSI/ISEA 107-2015 Type R Class 3.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty