Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mesh Overpants, PortWest

Supplier: Portwest
US386YERS/M US386YERXX/3X US386YERL/XL US386YER4X/5X
76106-132EA 24.84 USD InStock
76106-132 76106-130 76106-136 76106-134
Mesh Overpants, PortWest
Workwear
Mesh overpants are manufactured from 100% polyester mesh.

  • Silver reflective tape (2") with contrast material
  • Cooling mesh fabric
  • Elasticated ankles with hook and loop closure
  • Drawstring elasticated waistband

Mesh overpants feature two rear pockets and two side pass-through pockets. Machine wash warm/mild and line dry. Do not bleach, iron, or dry clean.

Certifications: ANSI/ISEA 107-2015 Supplemental Class E.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty