Would you like to log in again?
Hi-Vis Premium 2-in-1 Bomber Jackets, PortWest®
Supplier: Portwest
US364YBRM US364YBRL US364YBR5XL US364YBRXXL US364YBRXXXL US364YBRXL US364YBRS US364YBR4XL
76106-176EA 98.18 USD InStock
76106-176 76106-186 76106-174 76106-184 76106-178 76106-188 76106-182 76106-180
Hi-Vis Premium 2-in-1 Bomber Jackets, PortWest®
Workwear
Pack away adjustable hood, elasticated waistband and adjustable cuffs and detachable fleece liner with fleece sleeves for extra warmth.
  • With 2" silver reflective tape
  • Outer jacket pockets – one left chest radio, two lower front, one inner tablet, one sleeve
  • One inner chest inner jacket pocket
  • Two mic tabs
  • Waterproof with sealed seams
Contrast panel for protection against dirt.
Certifications: ANSI/ISEA 107-2010 Class 3:2. EN ISO 20471 Class 3.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty