Would you like to log in again?
Deep Vacuum Sublimation Apparatus, Ace Glass Incorporated
Supplier: Ace Glass
8020-07 8020-09 8020-05
76107-022EA 1502.57 USD InStock
76107-022 76107-024 76107-026
Deep Vacuum Sublimation Apparatus, Ace Glass Incorporated
Freeze Dryers
Improved sublimer designed for deep vacuum, with the vacuum valve on the top of the sublimer to keep the sublimate around the cold finger out of your vacuum trap
  • Features a Viton O-Ring and a quick release clamp for deep vacuum operation
  • Duran Flange is 150 mm
  • Complete item includes outer body, inner cold finger lid, and quick release clamp
  • Outer body and Inner cold finger lid also sold as separate items
Increased spacing between the cold finger and outer body leaves more room for your product.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty