Would you like to log in again?
Caframo H Base Support Stands, Ace Glass Incorporated
Supplier: Ace Glass
13568-04 13568-02 13568-06
76107-088EA 584.61 USD InStock
76107-088 76107-090 76107-092
Caframo H Base Support Stands, Ace Glass Incorporated
Support Stands
Caframo H-Base stands offer improved stability for benchtop reaction assemblies
  • Base is manufactured from cast alloy of zinc and aluminum, and coated with a chemical resistant epoxy paint
  • Primary support rod is manufactured from 1" O.D. 304 stainless steel tubing
  • Stands are available in three different heights
  • Stands are tapped for two additional 5/8" O.D. secondary support rods
  • Secondary supports are of 28" height and manufactured from 304 stainless steel rod
Clamp attaches to 1/2" to 1" O.D. support stand uprights, and supports up to 5/8" (16 mm) O.D. rod-mounted equipment.
Delivery information: Includes primary support rod with a plastic cap.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty