Would you like to log in again?
PTFE [ST] Joint Inserts, for Flat Stainless Steel Lids, Ace Glass Incorporated
Supplier: Ace Glass
6412-09 6412-13 6412-05 6412-11 6412-07
89427-916EA 328.09 USD InStock
89427-916 89427-912 89427-914 76111-086 89427-910
PTFE [ST] Joint Inserts, for Flat Stainless Steel Lids, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
Solid, virgin PTFE, standard taper joint inserts and Duran® flange inserts.
  • For use with stainless steel, flat heads
  • Bottom reverse thread on joint inserts includes PTFE collar and FETFE O-Ring
  • Top outer [ST] joint that matches glass [ST] inner joints
  • Flange insert matches glass Duran flange
  • O-Ring not included for Duran® flange insert
These joints are for use with stainless steel, flat heads. The top of the insert features an outer standard taper joint that matches glass standard taper inner joints. The bottom is made with a reverse thread and includes a PTFE collar and FETFE O-Ring for attaching to the flat heads. Top of insert is also machined to accept certain joint clips. For use with flat stainless steel lids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty