Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
DBI-SALA® ExoFit™ Vest-Style Positioning Harness with Back and Front D-Rings, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-1108583
76111-156EA 468.18 USD InStock
76111-156
DBI-SALA® ExoFit™ Vest-Style Positioning Harness with Back and Front D-Rings, ORS Nasco
Fall Protection Fall Harnesses
Lightweight, moisture wicking, breathable padding with soft edging wraps around the wearer for the ultimate in comfort, security, and no-tangle donning.
  • Back D-ring provides a safe connection for a fall arrest system, and keeps users upright every time
  • Hip D-rings are ideal for positioning, to hold and sustain users at a specific work location
  • Soft, lightweight webbing is extremely strong, yet isn't stiff or coarse for added comfort and durability
  • Built-in reflective piping enhances visibility in low light conditions
  • Parachute torso buckles complete the 5-point adjustment system for a perfect fit
Anti-tear ballistic nylon is abrasion-resistant for rugged strength and durability, and has twice the endurance of standard nylon or polyester. A Velcro® cover protects harness labels and holds an i-Safe™ identification tag for recording and storing information on inspections.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty