Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Protecta PRO-STOP™ Shock Absorbing Lanyards, ORS Nasco
Supplier: ORS Nasco
098-1340220
76111-172EA 69.94 USD InStock
76111-172
Protecta PRO-STOP™ Shock Absorbing Lanyards, ORS Nasco
Fall Protection Fall Lanyards and Ropes
  • Durable polyester webbing and an energy absorbing design limits fall arrest forces to 900 lbs. or less
  • 3,600 lb. gated hooks meet ANSI Z359 (USA only)
Certifications: Meets all applicable industry standards including OSHA and ANSI Z359.1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty