Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
DBI-SALA® Rope Lifeline with Snap Hooks, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-1202794
76111-338EA 166.54 USD InStock
76111-338
DBI-SALA® Rope Lifeline with Snap Hooks, ORS Nasco
Fall Protection Fall Lanyards and Ropes
Ideal for use as a vertical lifeline system with a rope grab.
  • Equipped with i-Safe™ RFID identification tag for recording and storing information on inspections
  • Durable, high strength 5-tuck splice provides an extra layer of ruggedness, strength, and field longevity
  • Polyester/polypropylene blend offers additional abrasion and UV resistance for added durability and longevity
Built-in self locking snap hook at one end offers fast and safe connection to an anchorage connector, such as a tie-off adaptor for added productivity.
Certifications: OSHA 1910.66, OSHA 1926.502, Capital Safety General Manufacturing Requirements, ANSI A10.32, ANSI Z359.1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty