Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
DBI-SALA® Force2™ Elastic 100% Tie-Off Shock Absorbing Lanyards, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-1246032
76111-376EA 550.84 USD InStock
76111-376
DBI-SALA® Force2™ Elastic 100% Tie-Off Shock Absorbing Lanyards, ORS Nasco
Fall Protection Fall Lanyards and Ropes
  • Stretches in reaction to wearer movements
  • Tech-Lite™ aluminum D-rings on each leg end for rescue
  • Abrasion-resistant polyester webbing
  • Energy-absorbing design limits fall arrest forces
  • User friendly self-locking snap hooks at center
  • Self-locking aluminum rebar hook at leg ends
Certifications: ANSI A10.32, ANSI Z359.13, ASTM F887, OSHA 1910.66, OSHA 1926.502.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty