Would you like to log in again?
DBI-SALA Nano-Lok™ Quick Connect Self Retracting Lifelines for Hot Work Use, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-3101497 098-3101491
76111-418EA 386.96 USD InStock
76111-418 76111-420
DBI-SALA Nano-Lok™ Quick Connect Self Retracting Lifelines for Hot Work Use, ORS Nasco
Fall Protection Fall Arresters
Designed to provide a snug fit to the worker's back, eliminating nuisance flopping.
  • Arc flashed to 8 cal/cm2 for hot work use in applications like welding, grinding and torching
  • Automatically locks and closes for added safety, and its rugged construction is built to last
  • Compact and durable housing is designed to withstand the demands of challenging jobsites
  • Swiveling anchorage loop limits lifeline twisting for smoother operation and added worker satisfaction
  • Quick activating speed sensing brake system stops a falling worker within inches, providing more protection at low heights
  • High capacity energy absorber and impact indicator protected by Kevlar® fiber cover
  • Built-in RFID identification tag helps the user record and store information on inspections
Built-in connector for single-leg SRL direct attachment to harness installs to the webbing of harness, leaving the dorsal D-ring open for other equipment or rescue. Kevlar® fiber webbing lifeline provides ultimate flame resistance and the retractable design provides freedom of movement, keeps the lifeline out of the worker's way, and reduces dragging, snagging, and trip falls.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty