Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
DBI-SALA® Rollgliss™ R550 Rescue and Descent Devices, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-3327100
76111-466EA 1816.89 USD InStock
76111-466
DBI-SALA® Rollgliss™ R550 Rescue and Descent Devices, ORS Nasco
Fall Protection Fall Protection Kits
Effective safety equipment for rescue, evacuation, or assisted rescues.
  • Light and compact design
  • Corrosion-resistant durable aluminum cast housing
  • Features fully automatic controlled descent
  • Integrated rope ears allow for greater descent speed control
  • Dual brakes provide additional safety and security
  • Super static kernmantle rope is lightweight, yet extremely strong and durable
Includes rescue wheel, 4' (1.2m) anchor sling, (2) carabiners, and carrying bag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty