Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
DBI-SALA® Sealed Blok™ Self-Retracting Lifelines, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-3400923 098-3400900
76111-476EA 6470.22 USD InStock
76111-476 76111-472
DBI-SALA® Sealed Blok™ Self-Retracting Lifelines, ORS Nasco
Fall Protection Fall Lanyards and Ropes
  • Features galvanized wire rope lifeline with self-locking swivel snap hook
  • Cast aluminum housing and heavy gauge steel end plates
  • Sealed design keeps the critical working components (brake, power spring, energy absorber) free of dirt, grease, water, and chemicals
Certifications: Meets or exceeds OSHA 1926.502, OSHA 1910.66, ANSI Z359.1, ANSI A10.32.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty