Would you like to log in again?
DBI-SALA® Rebel™ Self Retracting Cable Lifelines, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-3590550 098-3590016 098-3590500 098-3590590
76111-510EA 1433.43 USD InStock
76111-510 76111-502 76111-504 76111-506
DBI-SALA® Rebel™ Self Retracting Cable Lifelines, ORS Nasco
Fall Protection Fall Lanyards and Ropes
  • Abrasion-resistant and extremely durable lifeline for added field longevity
  • Retractable design provides freedom of movement and reduces dragging, snagging, and trip falls
  • Rugged, thermoplastic housing is compact and lightweight
  • Side lifeline exit with ergonomic handle enables smooth operation and reduces cable wear
Swiveling self-locking snap hook with impact indicator automatically locks and closes for added safety, swivels to limit lifeline twisting, and provides visual indication of fall arrest. Speed sensing, all-metal brake system stops a falling worker within inches and provides smooth consistent operation, allowing the user to maintain normal work pace.
Certifications: Meets all applicable industry standards including OSHA and ANSI Z359.1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty