Biomek® Pipette Tips from Beckman Coulter® are the only tips on the market that are designed, validated, and approved for use on Biomek® Liquid Handlers.
- Color-coded racks
- Easy visual checks
Tip definitions, pipetting techniques, and templates are built into Biomek® software. For best in total system performance, use only Beckman Coulter® validated pipette tips on Biomek® workstations.
Products that are designated as Sterile are either sterilized using Gamma radiation or ethylene oxide (EtO) gas in accordance with ANSI/AAMI/150 11135 or ANSI/AAMI/ISO 11137 guidelines. The sterilization processes certify a sterility assurance level of 10-6. Biomek® P30, P30XL, P20, P50, and P250 tips are EtO gas-sterilized for critical low-volume pipetting performance.
Tip racks are color-coded and matched in Biomek® software for easy-to-identify visual checks during method development and confirmation of the deck before a run.
Certifications: All pipette tips are certified to be free from DNase/RNase, DNA (Human and Mouse), PCR inhibition, pyrogen, endotoxins, and trace-metals.
Ordering information: AP384 tips are designed for use on the Biomek® AP384 Multichannel Head. AP96 Tips are designed for use on the Biomek® AP96 Multichannel head and Biomek® 3000 Pipetting Tools. Span-8 tips are designed for use on the Biomek® Span-8 pod. Biomek® 2000 and 3000 tips are designed for use on the respective pipetting tools.