Would you like to log in again?
Urea Assay Kit II, BioAssay Systems
Supplier: BioAssay Systems
DUR2-100
76121-348EA 246.84 USD InStock
76121-348
Urea Assay Kit II, BioAssay Systems
Assays Cellular Assays
For quantitative enzymatic determination of urea
  • Fast and sensitive. Linear detection range (20 μL sample): 1 mg/mL (0.17 mM) to 100 mg/mL (17 mM) urea in 96-well plate assay.
  • The procedure involves adding a single working reagent, and reading the absorbance after 5 minutes.
  • Room temperature assay. No 37°C heater is needed.
  • Homogeneous "mix-incubate-measure" type assay. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day.
  • Method: OD557nm
  • Samples: Urea in biological samples (e.g. plasma, serum, urine) and food/beverage samples (e.g. milk)
UREA, the major end product of protein catabolism in animals, is primarily produced in the liver and secreted by the kidneys. It is the primary vehicle for removal of toxic ammonia from the body. Urea determination is very useful for medical clinicians to assess kidney function of patients. In general, increased urea levels are associated with nephritis, renal ischemia, urinary tract obstruction, and certain extrarenal diseases (e.g. congestive heart failure, liver diseases, and diabetes). Decreased levels often indicate acute hepatic insufficiency, but may also result from over vigorous parenteral fluid therapy. BioAssay Systems' colorimetric urea (BUN) assay is based on urease catalyzed conversion of urea to ammonium and carbon dioxide. The pH of the reaction medium is monitored by a chromogen and the intensity of the reaction product at 557 nm is directly proportional to the urea concentration in the sample.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty