Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyvinylpyrrolidone

Supplier: BIOASSAY SYSTEMS
Synonyms: PolyvidonePolyvinylpyrrolidonPVP

PVPP-10
76121-356EA 34.3 USD InStock
76121-356
Polyvinylpyrrolidone
Polyvinylpyrrolidone
Polyvinylpyrrolidone (PVPP) used for preparation of wine samples for use with the QuantiQuik™ Histamine Quick Test Strips.

  • Powder


Formula: (C₆H₉NO)n
MDL Number: MFCD01076626
CAS Number: 9003-39-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty