Would you like to log in again?
G6PDH Inhibitor Screening Kit, BioAssay Systems
Supplier: BioAssay Systems
IGPDH-100
76121-360EA 559.52 USD InStock
76121-360
G6PDH Inhibitor Screening Kit, BioAssay Systems
Assays Cellular Assays
HTS for inhibitor screening and evaluation of glucose-6-phosphate dehydrogenase inhibitors
  • Non-radioactive assay.
  • Homogenous “mix-incubate-measure” type assay. Can be readily automated on HTS liquid handling system.
  • Rapid and reliable. Can be completed in 30 minutes and no 37°C heater is needed.
  • Method: OD565nm
  • Samples: Glucose-6-phosphate dehydrogenase
GLUCOSE-6-PHOSPHATE DEHYDROGENASE (G6PDH) is a cytosolic enzyme in the pentose phosphate pathway which supplies reducing energy to cells by maintaining the level of the co-enzyme nicotinamide adenine dinucleotide phosphate (NADPH). G6PDH reduces nicotinamide adenine dinucleotide phosphate (NADP) to NADPH while oxidizing glucose-6-phosphate (G6P). Humans with a genetic deficiency of G6PDH are predisposed to non-immune hemolytic anemia. Recently, studies have found G6PDH plays a critical role in survival, proliferation, and metastasis of cancer cells. Therefore, inhibitors of the enzyme are attractive candidates for new cancer therapeutics. BioAssay Systems' non-radioactive, colorimetric G6PDH assay is based on the reduction of the tetrazolium salt MTT in a NADPH-coupled enzymatic reaction to a reduced form of MTT which exhibits an absorption maximum at 565 nm. The increase in absorbance at 565 nm is proportional to the enzyme activity. The percent inhibition of a test compound can be determined by comparing the activity of G6PDH treated with a test compound to the activity of untreated G6PDH.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty