Would you like to log in again?
CRITERION™ HardyCHROM™ ECC Complete, Dehydrated Culture Media, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C9031
76121-388EA 874.84 USD InStock
76121-388
CRITERION™ HardyCHROM™ ECC Complete, Dehydrated Culture Media, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the detection, differentiation, and enumeration of <italic>E.coli</italic> and coliforms in food or water samples based on colony color.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty