Would you like to log in again?
COLUMN CW20M 4% 0.8% KOH CBKB NOC 2M 2MM
Supplier: Avantor
CarboBlack Column (packed) On CarboBlack B 2m^ CarboBlack Columns (packed) On CarboBlack B, 4%, 0.8% Koh Cbkb 60/80 Length: 2M, ID: 2Mm Noc, unique selectivity & very little adsorption for alcohol analyses, can hold up to a 10% loading of a nonsilicone li
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN CW20M 4% 0.8% KOH CBKB NOC 2M 2MM 76121-824   Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty