Would you like to log in again?
COLUMN MS5A SILCO 2M 2MM FOR TELEDYNE GC
Supplier: Avantor
Molecular Sieve Column 5A 80/100 2m^ Molecular Sieve Column 5A 80/100 Length: 2m, 2mm ID, Silco for Teledyne GCs, separate permanent gases at above-ambient temperatures, preactivated and ready to use,column comes with metal end-fitting to prevent water/ca
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN MS5A SILCO 2M 2MM FOR TELEDYNE GC 76121-840   Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty