Would you like to log in again?
GOOSENECK SPLITLESS LINER 4X6.3X78.5 MM
Supplier: Avantor
Gooseneck Splitless Liner^ Gooseneck Splitless Liner, IDxOD x Length: 4.0 mmx6.3 mm x 78.5 mm, for Agilent GCs, With deactivated wool, Applications: trace samples <2 uL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GOOSENECK SPLITLESS LINER 4X6.3X78.5 MM 76121-948   Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty