Would you like to log in again?
STRAIGHT INLET LINER 4.0MMX92.1MM X6.2MM
Supplier: Avantor
Straight Inlet Liner 92.1mm^ Straight Inlet Liner, ID:4.0 mm, Length:92.1 mm, OD:6.2 mm, Deactivation:Standard, Packing:Deactivated Wool, Material:Borosilicate Glass, for PerkinElmer Auto SYS and Clarus GCs equipped with split/splitless inlets
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STRAIGHT INLET LINER 4.0MMX92.1MM X6.2MM 76121-954   Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty