Would you like to log in again?
STRAIGHT INLET LINER 5MM 105 MM 8.0 MM
Supplier: Avantor
Straight Inlet Liner 5mm^ Straight Inlet Liner, ID: 5mm, Length:105 mm, OD:8.0 mm, Deactivation:Standard, Material:Borosilicate Glass, for Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
STRAIGHT INLET LINER 5MM 105 MM 8.0 MM 76121-960   Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty