Would you like to log in again?
VIAL HEAD SPACE CRIMP TOP AMB 20MM 20 ML
Supplier: MicroSolv Technology Corp
Vial Head Space 20mm 20ml^ Vial Head Space, 20mm, crimp top, Colour: Amber, tapered neck flat bottom, 75mm x 23mm, made of the highest quality borosilicate glass, identical to OEM specs and are offered in a full range of septa, Use with Agilent, Dani Fisons autosamplers, BASIK Brand, Size: 20ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIAL HEAD SPACE CRIMP TOP AMB 20MM 20 ML 76122-394 95025-E-AP20A Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty