Would you like to log in again?
VIAL HEAD SPACE CRIMP TOP AMB 20MM 27 ML
Supplier: MicroSolv Technology Corp
Vial Head Space 20mm 27 ml^ Vial Head Space, 20mm, crimp top, Colour: Amber,tapered neck flat bottom, 60mm x 30mm, made of the highest quality borosilicate glass, identical to OEM specs and are offered in a full range of septa, Use with Agilent, Dani Fisons autosamplers, BASIK Brand, Size: 27ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIAL HEAD SPACE CRIMP TOP AMB 20MM 27 ML 76122-396 95025-E-AP27C Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty