Would you like to log in again?
BioFit MVMT™ Critical Performance Seating, BioFit
Supplier: BioFit
MP-TL-T-AV126 MP-ML-T-AV126 MP-TM-T-AV126 MP-TH-T-AV126 MP-MH-T-AV126
76124-542EA 2812.45 USD InStock
76124-542 76124-532 76124-534 76124-536 76124-538
BioFit MVMT™ Critical Performance Seating, BioFit
Chairs
MVMT™ (Movement) ergonomic seating is changing the dynamic of the seated worker experience in today’s workplace, combining a forward-looking European perspective with the BioFit tradition of technological precision and proven durability.
  • Intuitive functionality including weight-activated control and synchronous seat and backrest
  • Engineered for task-specific range of motion and 24/7/365 use
  • Lumbar support and 3" adjustable backrest height
  • Aesthetic complements interior design trends, positively impacting corporate image
  • Optional easy wipe down to ISO Class 3 clean room models available
  • Exclusive QR code product tag delivers individual webpage for every MVMT unit manufactured
Weight activated Pro control includes 2.5" seat slide which provides functional seat depth from 16.4" to 18.9", pneumatic seat height adjustment, forward seat tilt, smooth and seamless forward backrest adjustment (a BioFit exclusive), and synchronous seat/back recline at 5:1 ratio
BioFit’s seating solutions are crafted with user comfort, health and wellbeing in mind. Products meet or exceed rigorous industry standard and carry a 13-years warranty with a lifetime warranty on the pneumatic mechanisms.
Over 70 years’ background improving the workplace experience to provide ergonomic, LEED® compliant seating for healthcare, technology, education, lab, industrial, office and custom applications.
Seat/back recline features an automatic weight-adjusted tension mechanism, in addition to manual tension adjustment for fine-tuning and a function to lock into one of four reclined angles.
Ordering information: Modular componentry permits numerous configuration alternatives tailored to suit specific tasks, body types, interior design trends and budgetary considerations in the following work spaces and beyond: Clean Room/Static Control/Laboratory/Healthcare/Industrial/Standing Workstation Applications. Contact your Avantor representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty