Would you like to log in again?
Quality Economy Brand Luer Lock Syringes, Air-Tite
Supplier: Air-Tite
ML60 ML3 ML30 ML20 ML1 ML10 ML5
76124-662BX 13.48 USD InStock
76124-662 76124-664 76124-666 76124-668 76124-658 76124-660 76124-670
Quality Economy Brand Luer Lock Syringes, Air-Tite
Syringes General-Purpose Syringes
Quality disposable syringes, made to exacting specifications. Great value at a low cost.
  • Latex-free
  • Non-toxic
  • Pyrogen-free
  • Positive plunger stop
Why spend more if you are looking for a basic, functional syringe? The Air-Tite brand syringes are a quality syringe, manufactured under ISO standards. Suitable for laboratory and veterinary use, these basic 3-part syringes offer a lubricated rubber tip plunger for smooth gliding in sterile, individually wrapped packaging.
From 1 to 60 ml in both luer slip and luer lock, these syringes are high-quality and economically priced to fit any budget.
Certifications: Meets ISO standards.
Packaging: Sterile, individually wrapped.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty