Would you like to log in again?
Oxford Listeria Supplement, Modified, HiMedia Laboratories
Supplier: HiMedia Laboratories
FD172-5VL
76124-812PK 91.13 USD InStock
76124-812
Oxford Listeria Supplement, Modified, HiMedia Laboratories
Media for Microbiology Additives and Supplements for Microbiology
An antimicrobial supplement recommended for selective isolation of Listeria species.
  • One vial sufficient for 500 ml medium
  • Store at 2 to 8 °C
Caution: Not for medicinal use.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty