Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Women's Stretch Oxford Shirts, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

FL254TKXL FL254WHXS FL254FBS FL254WHL FL254LBXL FL254BSL FL254FBL FL254TKM FL254WHM FL254BSS FL254BSXL FL254LBL FL254TK2X FL254WHXL FL254LB2X FL254TKXS FL254FBXS FL254LBM FL254TKL FL254TKS FL254BS2X FL254WHS FL254BSXS FL254LBS FL254BSM FL254LBXS FL254FBXL FL254FB2X FL254WH2X FL254FBM
76144-834EA 78.84 USD InStock
76144-834 76144-978 76144-832 76144-976 76145-120 76145-122 76145-124 76145-048 76145-126 76145-128 76145-108 76144-980 76144-842 76144-982 76144-840 76144-846 76144-844 76144-838 76145-050 76144-836 76145-052 76145-130 76145-054 76145-132 76145-110 76145-134 76145-112 76145-114 76145-116 76145-118
Women's Stretch Oxford Shirts, Dickies®
Workwear
Dickies® style FL254, long sleeve.

  • Mechanical stretch fabric for ease of movement
  • Tailored fit
  • Easy care stain release, moisture wicking, resists wrinkles
  • Pearlized buttons
  • Two-piece lined collar with collar stays
  • Back tuck pleats, left chest pocket
  • Tailored sleeve placket and cuff

Made of 5 oz. Oxford flex fabric, 60% cotton/40% polyester.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty