Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Women's Premium Flat Front Pants, Plus Sizes, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

FPW221DN22WUU FPW221DC20WUU FPW221DS22WUU FPW221BK24WUU FPW221BK18WUU FPW221DN20WUU FPW221DC22WUU FPW221DC20WRG FPW221DN24WUU FPW221BK24WRG FPW221BK22WUU FPW221DC24WRG FPW221DS18WUU FPW221BK20WRG FPW221DC18WRG FPW221DN22WRG FPW221BK20WUU FPW221DC22WRG FPW221BK22WRG FPW221DC24WUU FPW221DS20WUU FPW221DN18WRG FPW221DS24WUU FPW221DN24WRG FPW221DN18WUU FPW221DN20WRG FPW221BK18WRG FPW221DC18WUU
76145-978EA 94.62 USD InStock
76145-978 76145-814 76145-816 76145-818 76146-008 76146-006 76146-004 76146-002 76145-980 76145-982 76146-000 76145-984 76145-820 76145-986 76145-822 76145-988 76145-824 76145-826 76146-014 76146-012 76145-990 76146-010 76145-992 76145-994 76145-996 76145-998 76145-976 76145-812
Women's Premium Flat Front Pants, Plus Sizes, Dickies®
Workwear
Dickies® style FPW221, relaxed fit, straight leg.

  • Fade resistant
  • Resists wrinkles
  • Premium construction through rise for shape and comfort
  • Stretch interlining in waistband for expanded comfort waist
  • Sits slightly below waist
  • Right back welt pocket with button closure
  • Hook and eye waist closure

Made of 6.75 oz. flex twill, 65% polyester/35% cotton.

Sizing Chart: RG (Regular) size designations represent a 31.5" inseam. UU size designations represent a 37" inseam, unhemmed.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty