Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Women's Premium Curvy Pants, Plus Sizes, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

FPW441BK38WUU FPW441DN24WUU FPW441BK24WUU FPW441BK32WUU FPW441DN22WUU FPW441DN18WUU FPW441DS24WUU FPW441BK36WUU FPW441DS20WUU FPW441BK22WUU FPW441BK20WUU FPW441BK26WUU FPW441BK30WUU FPW441BK40WUU FPW441DN20WUU FPW441BK28WUU FPW441DS22WUU FPW441BK18WUU FPW441DS18WUU
76146-090EA 94.62 USD InStock
76146-090 76146-080 76146-108 76146-106 76146-118 76146-104 76146-116 76146-102 76146-114 76146-100 76146-112 76146-098 76146-110 76146-088 76146-086 76146-094 76146-084 76146-092 76146-082
Women's Premium Curvy Pants, Plus Sizes, Dickies®
Workwear
Dickies® style FPW441, curvy fit, straight leg.

  • For women with smaller waist and fuller hips through the seat
  • Resists wrinkles, easy care stain release, fade resistant
  • Premium continuous construction through rise for better shape and comfort
  • Contour waistband to prevent gapping, hook and eye closure
  • Two back welt pockets, E-Z Touch fabric
  • Plus size has extra darts for better fit
  • 6.75 oz. flex twill, 65% polyester/35% cotton

Sizing Chart: UU size designations represent a 37" inseam, unhemmed.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty