Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Insulated Color Block Jackets, Dickies®

Supplier: VF Imagewear

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

LJ605DNSV3X LJ605BKSV3X LJ605BKSV4X LJ605DNSV4X LJ605DNSVM LJ605BKSVM LJ605BKSV2X LJ605BKSV5X LJ605DNSVS LJ605BKSVS LJ605DNSV2X LJ605DNSVL LJ605BKSVXL LJ605DNSVXL LJ605BKSVL LJ605DNSV5X
76146-818EA 157.73 USD InStock
76146-818 76146-838 76146-828 76146-836 76146-848 76146-826 76146-834 76146-824 76146-846 76146-832 76146-822 76146-844 76146-830 76146-820 76146-842 76146-840
Insulated Color Block Jackets, Dickies®
Workwear
Dickies® style LJ605.

  • Water resistant, resists wrinkles
  • Full zip jacket, two front slash welt pockets
  • Contrasting color block detail on shoulders
  • Adjustable waist tabs, adjustable cuffs
  • Pencil pocket on left sleeve
  • 8 oz. twill, 65% polyester/35% cotton
  • Lining: 4 oz. nylon taffeta quilted to polyurethane foam

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty